(Pentamethylcyclopentadienyl)hafnium trichloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pentamethylcyclopentadienylhafnium(IV) trichloride

H27010-03 H27010-06
AAH27010-03EA 70.5 USD
AAH27010-03 AAH27010-06
(Pentamethylcyclopentadienyl)hafnium trichloride 98+%
(Pentamethylcyclopentadienyl)hafnium trichloride

Purity: 98+% CAS No.: 75181-08-7 Molecular Formula: C10H15Cl3Hf Formula Weight: 420.08 Melting Point: 170-172° UN No.: 3261 MDL No.: MFCD00070461

Formula: C₁₀H₁₅Cl₃Hf
MW: 420.07 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00070461
CAS Number: 75181-08-7
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR