(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1S,2S)-(+)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-TsDPEN, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide

H27006-06 H27006-14 H27006-03
AAH27006-06EA 517 USD
AAH27006-06 AAH27006-14 AAH27006-03
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide

CAS No.: 167316-27-0 Molecular Formula: C21H22N2O2S Formula Weight: 366.48 Melting Point: 127-130° Optical Rotation: +29° (c=1 in chloroform) MDL No.: MFCD03095684

Formula: C₂₁H₂₂N₂O₂S
MW: 366.48 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD03095684
CAS Number: 167316-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR