2-Hydroxy-5-pyrimidineboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26976-03
H26976-MD
AAH26976-03EA
871
USD
AAH26976-03
AAH26976-MD
2-Hydroxy-5-pyrimidineboronic acid pinacol ester 95%
2-Hydroxy-5-pyrimidineboronic acid pinacol ester
Purity: 95%
CAS No.: 1073354-84-3
Molecular Formula: C10H15BN2O3
Formula Weight: 222.05
Melting Point: 201-204°
MDL No.: MFCD09037497
Formula:
C₁₀H₁₅BN₂O₃ Melting Pt: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037497 CAS Number: 1073354-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...