Chlorobis(4-fluorophenyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Danger

H26953-03 H26953-06 H26953-MD
AAH26953-03EA 301 USD
AAH26953-03 AAH26953-06 AAH26953-MD
Chlorobis(4-fluorophenyl)phosphine 98%
Chlorobis(4-fluorophenyl)phosphine

Purity: 98% CAS No.: 23039-97-6 Molecular Formula: C12H8ClF2P Formula Weight: 256.62 UN No.: 3265 MDL No.: MFCD04972302

Formula: C₁₂H₈ClF₂P
MW: 256.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972302
CAS Number: 23039-97-6
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR