Ethyl-3-cyano-3-phenylpyruvate 97%

Supplier: Thermo Scientific Chemicals
Warning

H26951-14 H26951-06
AAH26951-14EA 113 USD
AAH26951-14 AAH26951-06
Ethyl-3-cyano-3-phenylpyruvate 97%
Ethyl-3-cyano-3-phenylpyruvate

Purity: 97% CAS No.: 6362-63-6 Molecular Formula: C12H11NO3 Formula Weight: 217.23 Melting Point: 125-129° MDL No.: MFCD00019801

Formula: C₁₂H₁₁NO₃
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00019801
CAS Number: 6362-63-6
EINECS: 228-844-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR