(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: rac-BIPHEN H2, 3,3'-Di-tert-butyl-5,5',6,6'-tetramethyl-[1,1'-biphenyl]-2,2'-diol, racemic-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol

H26949-06 H26949-03
AAH26949-06EA 101 USD
AAH26949-06 AAH26949-03
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 99%
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol

Purity: 99% CAS No.: 101203-31-0 Molecular Formula: C24H34O2 Formula Weight: 354.54 Melting Point: 164-169° MDL No.: MFCD01862439

Formula: C₂₄H₃₄O₂
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD01862439
CAS Number: 101203-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR