(R)-(-)-3-Fluoropyrrolidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26947-03
H26947-06
AAH26947-03EA
159
USD
AAH26947-03
AAH26947-06
AAH26947-MD
(R)-(-)-3-Fluoropyrrolidine hydrochloride 97%
(R)-(-)-3-Fluoropyrrolidine hydrochloride
Purity: 97%
CAS No.: 136725-55-8
Molecular Formula: C4H8FN.HCl
Formula Weight: 125.57
Melting Point: 181-184°
MDL No.: MFCD04038717
Formula:
C₄H₉ClFN Melting Pt: 181…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038717 CAS Number: 136725-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...