2-(3,4-Dimethoxyphenyl)ethanol 98%

Supplier: Thermo Scientific Chemicals

H26941-18 H26941-09
AAH26941-18EA 280 USD
AAH26941-18 AAH26941-09
2-(3,4-Dimethoxyphenyl)ethanol 98%
2-(3,4-Dimethoxyphenyl)ethanol

Purity: 98% CAS No.: 7417-21-2 Molecular Formula: C10H14O3 Formula Weight: 182.22 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 172-174°/17mm MDL No.: MFCD00002894

Formula: C₁₀H₁₄O₃
Boiling Pt: 172…174 °C/17 mm
Melting Pt: 45…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002894
CAS Number: 7417-21-2
EINECS: 231-032-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR