4'-Acetamido-2'-methylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-Acetyl-3-methylphenyl)acetamide

H26933-06 H26933-14
AAH26933-06EA 36.9 USD
AAH26933-06 AAH26933-14
4'-Acetamido-2'-methylacetophenone 97%
4'-Acetamido-2'-methylacetophenone

Purity: 97% CAS No.: 34956-31-5 Molecular Formula: C11H13NO2 Formula Weight: 191.23 Melting Point: 137-140° MDL No.: MFCD02258876

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD02258876
CAS Number: 34956-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR