Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

H26912-06 H26912-03
AAH26912-06EA 410 USD
AAH26912-06 AAH26912-03
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride 98+%
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride

Purity: 98+% CAS No.: 85959-83-7 Molecular Formula: C20H30Cl2Hf Formula Weight: 519.86 Melting Point: 300° MDL No.: MFCD00058887

Formula: C₂₀H₃₀Cl₂Hf
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00058887
CAS Number: 85959-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR