(S)-2-Amino-2-cyclohexylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Chg-OH, H-Cyclohexyl-Gly-OH, (S)-alpha-Aminocyclohexylacetic Acid

H26881-06 H26881-03
AAH26881-06EA 195 USD
AAH26881-06 AAH26881-03
(S)-2-Amino-2-cyclohexylacetic acid 98%
(S)-2-Amino-2-cyclohexylacetic acid

Purity: 98% CAS No.: 14328-51-9 Molecular Formula: C8H15NO2 Formula Weight: 157.22 Melting Point: 300° Optical Rotation: 31° (c=0.5 in 1N HCl) MDL No.: MFCD01311679

Formula: C₈H₁₅NO₂
MW: 157.2 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01311679
CAS Number: 14328-51-9

Order Now

Specification Test Results

Optical rotation +31° (c=0.5 in 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR