Trimethyl(vinyloxy)silane 97%

Supplier: Thermo Scientific Chemicals
Danger

H26876-03 H26876-06
AAH26876-03EA 31.5 USD
AAH26876-03 AAH26876-06
Trimethyl(vinyloxy)silane 97%
Trimethyl(vinyloxy)silane

Purity: 97% CAS No.: 6213-94-1 Molecular Formula: H2C=CHOSi(CH3)3 Formula Weight: 116.24 Flash Point: -19°(-2°F) Refractive Index: 1.389 Boiling Point: 74° UN No.: 1993 MDL No.: MFCD00054764

Formula: C₅H₁₂OSi
MW: 116.24 g/mol
Boiling Pt: 74 °C
Density: 0.779
Flash Pt: -19 °C(-2 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054764
CAS Number: 6213-94-1
EINECS: 228-281-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR