3,5-Dimethyl-4-hydroxybenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile

H26872-06 H26872-14
AAH26872-03EA 43.62 USD
AAH26872-03 AAH26872-06 AAH26872-14
3,5-Dimethyl-4-hydroxybenzonitrile 98%
3,5-Dimethyl-4-hydroxybenzonitrile

Purity: 98% CAS No.: 4198-90-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 123-127° MDL No.: MFCD00221716

Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00221716
CAS Number: 4198-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR