Cumylamine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: alpha,alpha-Dimethylbenzylamine, Phenylisopropylamine, alpha-Aminoisopropylbenzene, 2-Phenylpropan-2-amine

H26845-03 H26845-06
AAH26845-03EA 200 USD
AAH26845-03 AAH26845-06
Cumylamine 96%
Cumylamine

Purity: 96% CAS No.: 585-32-0 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 72°(162°F) Refractive Index: 1.5200 Odor: Amine-like Boiling Point: 197° UN No.: 2735 MDL No.: MFCD00134680

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 197 °C
Density: 0.94
Flash Pt: 72 °C(162 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134680
CAS Number: 585-32-0
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR