2-Formyl-6-picoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

H26841-14 H26841-06
AAH26841-14EA 510 USD
AAH26841-14 AAH26841-06
2-Formyl-6-picoline 98%
2-Formyl-6-picoline

Purity: 98% CAS No.: 1122-72-1 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 68°(155°F) Refractive Index: 1.527 Melting Point: 29-33° Boiling Point: 105-108°/12mm Beilstein Reference: 107476 MDL No.: MFCD00006291

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 105…108 °C/12 mm
Melting Pt: 29…33 °C
Flash Pt: 68 °C(155 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006291
CAS Number: 1122-72-1
EINECS: 214-359-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR