3,5-Bis(trifluoromethyl)thiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H26809-03 H26809-MD
AAH26809-03EA 140 USD
AAH26809-03 AAH26809-MD
3,5-Bis(trifluoromethyl)thiobenzamide 97%
3,5-Bis(trifluoromethyl)thiobenzamide

Purity: 97% CAS No.: 317319-15-6 Molecular Formula: C9H5F6NS Formula Weight: 273.20 Melting Point: 112-115° MDL No.: MFCD00278692

Formula: C₉H₅F₆NS
MW: 273.20 g/mol
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00278692
CAS Number: 317319-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR