6-Methyl-2-pyridinemethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxymethyl-6-methylpyridine, 6-Hydroxymethyl-2-picoline
H26794-06
H26794-14
AAH26794-06EA
87.3
USD
AAH26794-06
AAH26794-03
AAH26794-14
6-Methyl-2-pyridinemethanol 98%
6-Methyl-2-pyridinemethanol
Purity: 98%
CAS No.: 1122-71-0
Molecular Formula: C7H9NO
Formula Weight: 123.15
Flash Point: 109°(228°F)
Melting Point: 32-34°
Boiling Point: 105-108°/12mm
MDL No.: MFCD00023520
Formula:
C₇H₉NO MW: 123.16 g/mol Boiling Pt: 105…108 °C/12 mm Melting Pt: 32…34 °C Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023520 CAS Number: 1122-71-0 EINECS: 214-358-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...