3,5-Difluoro-4-methoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

H26778-03 H26778-06
AAH26778-03EA 184 USD
AAH26778-03 AAH26778-06
3,5-Difluoro-4-methoxyaniline 97%
3,5-Difluoro-4-methoxyaniline

Purity: 97% CAS No.: 363-47-3 Molecular Formula: C7H7F2NO Formula Weight: 159.14 Odor: Characteristic Melting Point: 77-80° UN No.: 2811 MDL No.: MFCD04115910

Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD04115910
CAS Number: 363-47-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR