Di(N-succinimidyl) carbonate ≥85% remainder N-Hydroxysuccinimide, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
H26629-03
H26629-14
H26629-06
AAH26629-03EA
32.3
USD
AAH26629-03
AAH26629-14
AAH26629-06
Di(N-succinimidyl) carbonate ≥85% remainder N-Hydroxysuccinimide, tech.
Di(N-succinimidyl) carbonate
Purity: 85%
CAS No.: 74124-79-1
Molecular Formula: C9H8N2O7
Formula Weight: 256.17
Melting Point: 190° dec.
Beilstein Reference: 1499137
MDL No.: MFCD00009767
Formula:
C₉H₈N₂O₇ MW: 256.17 g/mol Melting Pt: 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00009767 CAS Number: 74124-79-1 EINECS: 277-730-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...