2-(p-Tolyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H26574-06 H26574-14
AAH26574-06EA 56.1 USD
AAH26574-06 AAH26574-14
2-(p-Tolyl)pyridine 98%
2-(p-Tolyl)pyridine

Purity: 98% CAS No.: 4467-06-5 Molecular Formula: C12H11N Formula Weight: 169.22 Flash Point: 110°(230°F) Refractive Index: 1.617 Boiling Point: 170-180/20mm MDL No.: MFCD00006281

Formula: C₁₂H₁₁N
MW: 169.22 g/mol
Boiling Pt: 170…180/20 mm
Density: 0.99
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006281
CAS Number: 4467-06-5
EINECS: 224-734-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR