Methyl-2-amino-2-methylpropanoate hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl alpha-aminoisobutyrate hydrochloride, H-Aib-OMe.HCl, Methyl 2-aminoisobutyrate hydrochloride, H-Aib-OMe·HCl
H26550-03
H26550-06
AAH26550-03EA
64.4
USD
AAH26550-03
AAH26550-06
Methyl-2-amino-2-methylpropanoate hydrochloride 99%
Methyl-2-amino-2-methylpropanoate hydrochloride
Purity: 99%
CAS No.: 15028-41-8
Molecular Formula: C5H11NO2.HCl
Formula Weight: 153.61
Storage Temperature: 2°C to 8°C
Melting Point: ca 185° dec.
MDL No.: MFCD00214247
Formula:
C₅H₁₁NO₂·HCl Melting Pt: ∼ 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00214247 CAS Number: 15028-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...