1-Benzyl-1,4,7,10-tetraazacyclododecane
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Benzyl-1,4,7,10-tetraazacyclododecane
H26525-03
H26525-MD
AAH26525-03EA
458.77
USD
AAH26525-03
AAH26525-MD
1-Benzyl-1,4,7,10-tetraazacyclododecane
1-Benzyl-1,4,7,10-tetraazacyclododecane
CAS No.: 112193-83-6
Molecular Formula: C15H26N4
Formula Weight: 262.40
Melting Point: 84-86°
MDL No.: MFCD09263310
Formula:
C₁₅H₂₆N₄ MW: 262.401 g/mol Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD09263310 CAS Number: 112193-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...