5-Carbethoxyuracil 98%
Supplier: Thermo Scientific Chemicals
H26507-03
H26507-06
AAH26507-03EA
24.5
USD
AAH26507-03
AAH26507-06
5-Carbethoxyuracil 98%
5-Carbethoxyuracil
Purity: 98%
CAS No.: 28485-17-8
Molecular Formula: C7H8N2O4
Formula Weight: 184.15
MDL No.: MFCD00057337
Formula:
C₇H₈N₂O₄ MW: 184.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057337 CAS Number: 28485-17-8 EINECS: 249-053-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...