N-Boc-L-prolinal 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1-Boc-2-Formylpyrrolidine
H26495-06
H26495-03
AAH26495-06EA
448
USD
AAH26495-06
AAH26495-03
N-Boc-L-prolinal 96%
N-Boc-L-prolinal
Purity: 96%
CAS No.: 69610-41-9
Molecular Formula: C10H17NO3
Formula Weight: 199.25
Storage Temperature: -30°C to -10°C
Flash Point:
Formula:
C₁₀H₁₇NO₃ Boiling Pt: 228 °C Density: 1.06 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00274186 CAS Number: 69610-41-9 |
Specification Test Results
Optical rotation | -95° (c=1, chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...