3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphate, DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate
H26481-06
H26481-14
AAH26481-06EA
62.9
USD
AAH26481-06
AAH26481-14
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Purity: 98%
CAS No.: 165534-43-0
Molecular Formula: C11H14N3O5P
Formula Weight: 299.22
Storage Temperature: 2°C to 8°C
Melting Point: 71-76°
MDL No.: MFCD01236967
Formula:
C₁₁H₁₄N₃O₅P MW: 299.22 g/mol Melting Pt: 71…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD01236967 CAS Number: 165534-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...