2,4-Dichloro-5-fluorobenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26464-03
H26464-09
AAH26464-03EA
66.1
USD
AAH26464-03
AAH26464-09
2,4-Dichloro-5-fluorobenzonitrile 97%
2,4-Dichloro-5-fluorobenzonitrile
Purity: 97%
CAS No.: 128593-93-1
Molecular Formula: C7H2Cl2FN
Formula Weight: 190.00
Melting Point: 44-47°
UN No.: 3439
MDL No.: MFCD03840469
Formula:
C₇H₂Cl₂FN MW: 190.00 g/mol Melting Pt: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840469 CAS Number: 128593-93-1 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...