4-(Ethoxycarbonyl)phenylboronic acid 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid
H26356-03
H26356-06
AAH26356-03EA
32.3
USD
AAH26356-03
AAH26356-06
4-(Ethoxycarbonyl)phenylboronic acid 94%
4-(Ethoxycarbonyl)phenylboronic acid
Purity: 94%
CAS No.: 4334-88-7
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 137-140°
MDL No.: MFCD02179441
Formula:
C₉H₁₁BO₄ MW: 193.99 g/mol Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179441 CAS Number: 4334-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...