Diethyl-2,5-dibromoadipate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl 2,5-dibromohexanedioate, 2,5-Dibromoadipic acid diethyl ester

H26347-06 H26347-14
AAH26347-06EA 56.61 USD
AAH26347-06 AAH26347-14
Diethyl-2,5-dibromoadipate 98%
Diethyl-2,5-dibromoadipate

Purity: 98% CAS No.: 869-10-3 Molecular Formula: C10H16Br2O4 Formula Weight: 360.04 Melting Point: 64-67° UN No.: 3261 MDL No.: MFCD00075379

Formula: C₁₀H₁₆Br₂O₄
MW: 360.04 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00075379
CAS Number: 869-10-3
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR