1-Phenylethyl mercaptan 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26345-03
H26345-09
AAH26345-03EA
50.24
USD
AAH26345-03
AAH26345-09
1-Phenylethyl mercaptan 98%
1-Phenylethyl mercaptan
Purity: 98%
CAS No.: 6263-65-6
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 76°(169°F)
Refractive Index: 1.5580
UN No.: 2810
MDL No.: MFCD00053515
Formula:
C₈H₁₀S MW: 138.23 Density: 1.021 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053515 CAS Number: 6263-65-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...