Dimethyl (4-fluorobenzyl)malonate 97%

Supplier: Thermo Scientific Chemicals

H26310-06 H26310-03
AAH26310-06EA 137 USD
AAH26310-06 AAH26310-03
Dimethyl (4-fluorobenzyl)malonate 97%
Dimethyl (4-fluorobenzyl)malonate

Purity: 97% CAS No.: 252938-22-0 Molecular Formula: C12H13FO4 Formula Weight: 240.23 MDL No.: MFCD01934558

Formula: C₁₂H₁₃FO₄
MW: 240.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01934558
CAS Number: 252938-22-0
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR