6-Methylpicolinic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid
H26302-03
H26302-06
AAH26302-03EA
58.7
USD
AAH26302-03
AAH26302-06
6-Methylpicolinic acid 95%
6-Methylpicolinic acid
Purity: 95%
CAS No.: 934-60-1
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Odor: Odorless
Melting Point: 127-131°
Boiling Point: 100°/4.5mm
MDL No.: MFCD00023481
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 100 °C/4.5 mm Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023481 CAS Number: 934-60-1 EINECS: 213-287-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...