Cyclopropylboronic acid tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26254-03
H26254-06
AAH26254-03EA
95.4
USD
AAH26254-03
AAH26254-06
Cyclopropylboronic acid tech. 85%, Technical Grade
Cyclopropylboronic acid
Purity: 85%
CAS No.: 411235-57-9
Molecular Formula: C3H7BO2
Formula Weight: 85.90
Storage Temperature: -30°C to -10°C
Melting Point: 90-95°
MDL No.: MFCD04038750
Formula:
C₃H₇BO₂ MW: 85.90 g/mol Melting Pt: 90…95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038750 CAS Number: 411235-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...