2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP
H26226-06
H26226-22
H26226-14
AAH26226-06EA
149
USD
AAH26226-06
AAH26226-22
AAH26226-14
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Purity: 98%
CAS No.: 98327-87-8
Molecular Formula: C44H32P2
Formula Weight: 622.69
Melting Point: 281-285°
Beilstein Reference: 5321443
MDL No.: MFCD00010805
Formula:
C₄₄H₃₂P₂ Melting Pt: 281…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010805 CAS Number: 98327-87-8 EINECS: 619-338-0 Merck Index: 14,1223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...