tert-Butyl hydrogen malonate 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Tert-butoxy-3-oxopropanoic acid, mono-tert-Butyl malonate

H26223-06 H26223-14
AAH26223-06EA 57.9 USD
AAH26223-06 AAH26223-14
tert-Butyl hydrogen malonate 96%
tert-Butyl hydrogen malonate

Purity: 96% CAS No.: 40052-13-9 Molecular Formula: C7H12O4 Formula Weight: 160.17 Flash Point: 91°(195°F) Refractive Index: 1.426 Melting Point: 19-20° Boiling Point: 90°/2mm Beilstein Reference: 1765457 MDL No.: MFCD00191886

Formula: C₇H₁₂O₄
Boiling Pt: 90 °C/2 mm
Melting Pt: 19…20 °C
Density: 1.04
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191886
CAS Number: 40052-13-9
UN: 2922
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR