2-Bromo-4-nitro(trifluoromethoxy)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-4-nitro-1-(trifluoromethoxy)benzene
H26173-06
H26173-03
AAH26173-06EA
87
USD
AAH26173-06
AAH26173-03
2-Bromo-4-nitro(trifluoromethoxy)benzene 98%
2-Bromo-4-nitro(trifluoromethoxy)benzene
Purity: 98%
CAS No.: 200958-40-3
Molecular Formula: C7H3BrF3NO3
Formula Weight: 286.00
Refractive Index: 1.508
Formula:
C₇H₃BrF₃NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD04973758 CAS Number: 200958-40-3 EINECS: None |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...