4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26154-03
H26154-MD
AAH26154-03EA
213
USD
AAH26154-03
AAH26154-MD
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Purity: 97%
CAS No.: 878572-17-9
Molecular Formula: C8H3F4NO4
Formula Weight: 253.11
Melting Point: 120-124°
MDL No.: MFCD08282778
Formula:
C₈H₃F₄NO₄ MW: 253.11 g/mol Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD08282778 CAS Number: 878572-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...