HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
H26082-06
H26082-03
H26082-14
AAH26082-06EA
130.5
USD
AAH26082-06
AAH26082-03
AAH26082-14
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 99%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Purity: 99%
CAS No.: 148893-10-1
Molecular Formula: C10H15F6N6OP
Formula Weight: 380.23
Melting Point: ca 185° dec.
MDL No.: MFCD00274639
Formula:
C₁₀H₁₅F₆N₆OP MW: 380.2 g/mol Melting Pt: ca 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00274639 CAS Number: 148893-10-1 UN: 1325 ADR: "4.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...