Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate, Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate

H26079-14 H26079-06
AAH26079-14EA 700 USD
AAH26079-14 AAH26079-03 AAH26079-06
Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate 98%
Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate

Purity: 98% CAS No.: 23286-70-6 Molecular Formula: C7H11N3O2 Formula Weight: 169.18 Melting Point: 108° MDL No.: MFCD01210846

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
Melting Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD01210846
CAS Number: 23286-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR