3,6-Dimethyl-4-hydroxy-2-pyrone 97%

Supplier: Thermo Scientific Chemicals
Warning

H26074-03 H26074-06
AAH26074-03EA 152 USD
AAH26074-03 AAH26074-06
3,6-Dimethyl-4-hydroxy-2-pyrone 97%
3,6-Dimethyl-4-hydroxy-2-pyrone

Purity: 97% CAS No.: 5192-62-1 Molecular Formula: C7H8O3 Formula Weight: 140.14 Melting Point: 209-211° MDL No.: MFCD00090424

Formula: C₇H₈O₃
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00090424
CAS Number: 5192-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR