2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(2-(1,3-Dioxolan-2-yl)ethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H26071-03
H26071-06
AAH26071-03EA
68
USD
AAH26071-03
AAH26071-06
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester 97%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester
Purity: 97%
Molecular Formula: C11H21BO4
Formula Weight: 228.10
Boiling Point: 100°/1.5mm
MDL No.: MFCD03788722
Formula:
C₁₁H₂₁BO₄ MW: 228.10 g/mol Boiling Pt: 100 °C/1.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD03788722 CAS Number: 1073354-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...