4-Cyanophenylacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Cyanophenyl)acetic acid

H26062-03 H26062-06
AAH26062-03EA 127 USD
AAH26062-03 AAH26062-06
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid

Purity: 97% CAS No.: 5462-71-5 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 150-152° UN No.: 3439 MDL No.: MFCD06798066

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06798066
CAS Number: 5462-71-5
EINECS: 226-753-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR