N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (R,R)-TsDPEN, (1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine

H26061-06 H26061-03 H26061-14
AAH26061-06EA 392 USD
AAH26061-06 AAH26061-03 AAH26061-14
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥98%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide

Purity: 98+% CAS No.: 144222-34-4 Molecular Formula: C21H22N2O2S Formula Weight: 366.48 Melting Point: 127-132° Optical Rotation: -31° (c=1 in chloroform) MDL No.: MFCD02093428

Formula: C₂₁H₂₂N₂O₂S
MW: 366.48 g/mol
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD02093428
CAS Number: 144222-34-4

Order Now

Specification Test Results

Optical rotation -31° (c=1 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR