1-(Trifluoromethyl)cyclopentanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26055-03 H26055-MD
AAH26055-03EA 170 USD
AAH26055-03 AAH26055-MD
1-(Trifluoromethyl)cyclopentanecarboxylic acid 97%
1-(Trifluoromethyl)cyclopentanecarboxylic acid

Purity: 97% CAS No.: 277756-44-2 Molecular Formula: C7H9F3O2 Formula Weight: 182.15 Melting Point: 36-38° MDL No.: MFCD08445821

Formula: C₇H₉F₃O₂
MW: 182.15
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD08445821
CAS Number: 277756-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR