3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3,3-Trifluoro-2,2-dimethylpropanoic acid
H26053-03
H26053-MD
AAH26053-03EA
317
USD
AAH26053-03
AAH26053-MD
3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%
3,3,3-Trifluoro-2,2-dimethylpropionic acid
Purity: 97%
CAS No.: 889940-13-0
Molecular Formula: C5H7F3O2
Formula Weight: 156.12
Melting Point: 66-73°
Boiling Point: 76-77°/14mm
UN No.: 3261
MDL No.: MFCD08445819
Formula:
C₅H₇F₃O₂ Boiling Pt: 76…77 °C/14 mm Melting Pt: 66…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD08445819 CAS Number: 889940-13-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...