1-Pyridin-3-ylethylamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(1-Aminoethyl)pyridine, 1-(Pyridin-3-yl)ethanamine
H26047-03
H26047-MD
AAH26047-03EA
418
USD
AAH26047-03
AAH26047-MD
1-Pyridin-3-ylethylamine 96%
1-Pyridin-3-ylethylamine
Purity: 96%
CAS No.: 56129-55-6
Molecular Formula: C7H10N2
Formula Weight: 122.17
UN No.: 2735
MDL No.: MFCD01691641
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01691641 CAS Number: 56129-55-6 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...