4'-Fluoro-2-methylpropiophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Fluoroisobutyrophenone, 1-(4-Fluorophenyl)-2-methylpropan-1-one

H26036-06 H26036-03
AAH26036-06EA 445 USD
AAH26036-06 AAH26036-03
4'-Fluoro-2-methylpropiophenone 97%
4'-Fluoro-2-methylpropiophenone

Purity: 97% CAS No.: 26393-91-9 Molecular Formula: C10H11FO Formula Weight: 166.19 Refractive Index: 1.4980

Formula: C₁₀H₁₁FO
MW: 166.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038305
CAS Number: 26393-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR