4-(4-Fluorophenyl)butyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4-Fluorophenyl)butyric Acid
H26035-03
H26035-MD
AAH26035-03EA
76.1
USD
AAH26035-03
AAH26035-MD
4-(4-Fluorophenyl)butyric acid 97%
4-(4-Fluorophenyl)butyric acid
Purity: 97%
CAS No.: 589-06-0
Molecular Formula: C10H11FO2
Formula Weight: 182.20
Melting Point: 44-46°
MDL No.: MFCD03788503
Formula:
C₁₀H₁₁FO₂ MW: 182.2 g/mol Melting Pt: 44…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788503 CAS Number: 589-06-0 EINECS: 209-631-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...