2-Chloro-6-methylbenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26023-03
H26023-06
AAH26023-03EA
75.3
USD
AAH26023-03
AAH26023-06
2-Chloro-6-methylbenzaldehyde 98%
2-Chloro-6-methylbenzaldehyde
Purity: 98%
CAS No.: 1194-64-5
Molecular Formula: C8H7ClO
Formula Weight: 154.59
Flash Point: 99°(210°F)
Melting Point: 36-38°
Boiling Point: 74°/0.4mm
MDL No.: MFCD01934409
Formula:
C₈H₇ClO Boiling Pt: 74 °C/0.4 mm Melting Pt: 36…38 °C Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01934409 CAS Number: 1194-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...