4-tert-Butoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydroquinone Mono-tert-butyl Ether

H26013-03 H26013-06
AAH26013-03EA 86 USD
AAH26013-03 AAH26013-06
4-tert-Butoxyphenol 98%
4-tert-Butoxyphenol

Purity: 98% CAS No.: 2460-87-9 Molecular Formula: C10H14O2 Formula Weight: 166.22 Odor: Phenol-like Melting Point: 151-153° MDL No.: MFCD02183556

Formula: C₁₀H₁₄O₂
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD02183556
CAS Number: 2460-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR