6-Amino-3-cyano-2-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H26000-03 H26000-06
AAH26000-03EA 60.4 USD
AAH26000-03 AAH26000-06
6-Amino-3-cyano-2-methylpyridine 97%
6-Amino-3-cyano-2-methylpyridine

Purity: 97% CAS No.: 183428-90-2 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 164-166° UN No.: 3439 MDL No.: MFCD05865116

Formula: C₇H₇N₃
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD05865116
CAS Number: 183428-90-2
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR